Please login to your online account to display your discounted pricing

Zinc telluride, 99.998% (metals basis), Alfa Aesar™

Chemical Identifiers

CAS 1315-11-3
Physical Form 3 to 12mm Lump
Assay Percent Range 99.998% metal basis
Molecular Formula ZnTe
Formula Weight 192.97
MDL Number MFCD00043079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3659509

  SDS
alfa aesar
3659509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3659518

  SDS
alfa aesar
3659518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form 3 to 12mm Lump
Formula Weight 192.97
Quantity 10g
Color Red
Molecular Formula ZnTe
Assay Percent Range 99.998% metal basis
MDL Number MFCD00043079
CAS 1315-11-3
Odor Odorless