missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc telluride, 99.99% (metals basis)

$64.15 - $288.38

Chemical Identifiers

CAS 1315-11-3
Molecular Formula TeZn
Molecular Weight (g/mol) 192.98
MDL Number MFCD00043079
InChI Key NSRBDSZKIKAZHT-UHFFFAOYSA-N
Synonym ir8eb6g3vq
PubChem CID 3362486
IUPAC Name tellanylidenezinc
SMILES [Zn]=[Te]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4441206 View Documents Alfa Aesar™
4441206
5g Each for $64.15
 
AA4441214 View Documents Alfa Aesar™
4441214
25g Each for $288.38
 
Specifications

Chemical Identifiers

1315-11-3
192.98
NSRBDSZKIKAZHT-UHFFFAOYSA-N
3362486
[Zn]=[Te]
TeZn
MFCD00043079
ir8eb6g3vq
tellanylidenezinc

Specifications

1315-11-3
MFCD00043079
ir8eb6g3vq
[Zn]=[Te]
192.98
192.97
99.99%
−140 Mesh
3.56
1240°C
Zinc telluride
TeZn
14,10162
NSRBDSZKIKAZHT-UHFFFAOYSA-N
tellanylidenezinc
3362486
Powder
5.9g/mL
Odorless
Red
5g
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA