missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc telluride, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4441214

 View more versions of this product

Catalog No. AA4441214



Specifications

Specifications

Zinc telluride
(metals basis)
MFCD00043079
ir8eb6g3vq
NSRBDSZKIKAZHT-UHFFFAOYSA-N
tellanylidenezinc
3362486
-140 Mesh Powder
5.9 g/mL
Odorless
Red
25g
1315-11-3
TeZn
14,10162
It decomposes in water.
[Zn]=[Te]
192.98
192.97
99.99%
−140 Mesh
3.56
1240°C

Chemical Identifiers

1315-11-3
192.98
NSRBDSZKIKAZHT-UHFFFAOYSA-N
3362486
[Zn]=[Te]
TeZn
MFCD00043079
ir8eb6g3vq
tellanylidenezinc
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 215-260-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents