Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sulfite dihydrate, 98%

$63.73 - $194.57

Chemical Identifiers

Chemical Name or Material Zinc sulfite dihydrate
CAS 7488-52-0
Molecular Formula ZnSO3·2H2O
Formula Weight 181.46 (145.44 Anhydrous)
MDL Number MFCD00171738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1122722

 
alfa aesar™
A1122722
100g Each for $63.73

AAA1122736

 
alfa aesar™
A1122736
500g Each for $194.57
Description & Specifications

Specifications

Formula Weight 181.46 (145.44 Anhydrous)
Chemical Name or Material Zinc sulfite dihydrate
Assay 98%
Molecular Formula ZnSO3·2H2O
MDL Number MFCD00171738
CAS 7488-52-0
Quantity 100g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.