Learn More
Zinc sulfide, 99.99%, (trace metal basis), < 10 microns
CAS: 1314-98-3 | SZn | 97.44 g/mol
$183.05 - $183.05
Chemical Identifiers
| CAS | 1314-98-3 |
|---|---|
| Molecular Formula | SZn |
| Molecular Weight (g/mol) | 97.44 |
| MDL Number | MFCD00011301 |
| InChI Key | DRDVZXDWVBGGMH-UHFFFAOYSA-N |
| Synonym | zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide |
| PubChem CID | 14821 |
| SMILES | [S--].[Zn++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC223651000
|
Thermo Scientific Chemicals
223651000 |
100 g | Glass bottle |
Each for $183.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 1314-98-3 | |
| 97.44 | |
| DRDVZXDWVBGGMH-UHFFFAOYSA-N | |
| 14821 |
| SZn | |
| MFCD00011301 | |
| zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide | |
| [S--].[Zn++] |
Specifications
| 1314-98-3 | |
| Powder | |
| 99.99% | |
| ZnS | |
| 15, 10360 | |
| Solubility in water: insoluble. Other solubilities: soluble in acids | |
| [S--].[Zn++] | |
| 97.44 | |
| 97.44 | |
| Trace Metal Basis | |
| Zinc sulfide, 99.99%, <10 microns |
| White to Yellow | |
| 100 g | |
| SZn | |
| MFCD00011301 | |
| zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide | |
| DRDVZXDWVBGGMH-UHFFFAOYSA-N | |
| zinc(2+) sulfanediide | |
| 14821 | |
| 99.99% | |
| Glass bottle |
Safety and Handling
GHS H Statement
Contact with acids liberates toxic gas.
GHS P Statement:
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MOISTURE SENSITIVE
EINECSNumber : 215-251-3
RTECSNumber : ZH5400000
TSCA : TSCA
RUO – Research Use Only