missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sulfide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 13239A1

 View more versions of this product

Catalog No. AA13239A1



Specifications

Specifications

Zinc sulfide
(metals basis)
ZnS
14,10160
Soluble in acids. insoluble in water.
S=[Zn]
97.44
97.43
99.99%
Odorless
White
1kg
1314-98-3
SZn
MFCD00011301
albalith
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
sulfanylidenezinc
14821
2-20mm Lump
4.10 g/mL
2 to 20mm
1700°C

Chemical Identifiers

1314-98-3
97.44
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
14821
S=[Zn]
SZn
MFCD00011301
albalith
sulfanylidenezinc
Safety and Handling

Safety and Handling

EINECSNumber : 215-251-3

RTECSNumber : ZH5400000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents