missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sulfide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1323918

 View more versions of this product

Catalog No. AA1323918



Specifications

Specifications

Zinc sulfide
(metals basis)
ZnS
14,10160
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
sulfanylidenezinc
14821
Lumps
4.10g/mL
2 to 20mm
1700°C
1314-98-3
SZn
MFCD00011301
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
S=[Zn]
97.44
97.43
99.99%
Odorless
White
50g

Chemical Identifiers

1314-98-3
97.44
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
14821
S=[Zn]
SZn
MFCD00011301
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
sulfanylidenezinc
Safety and Handling

Safety and Handling

RTECSNumber : ZH5400000

TSCA : TSCA

Documents