missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Sulfide, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8829909

 View more versions of this product

Catalog No. AA8829909



Specifications

Specifications

Zinc Sulfide
99.9%, metal basis
MFCD00011301
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
sulfanylidenezinc
14821
Pieces, 3-12mm (0.1-0.5 in.)
1314-98-3
SZn
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
S=[Zn]
97.44
97.43
10g

Chemical Identifiers

1314-98-3
97.44
WGPCGCOKHWGKJJ-UHFFFAOYSA-N
14821
S=[Zn]
SZn
MFCD00011301
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
sulfanylidenezinc
SDS Documents