Please login to your online account to display your discounted pricing

Zinc Sulfide, 99.9% (Metal Basis), Alfa Aesar™

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material Zinc Sulfide
CAS 1314-98-3
Physical Form Pieces, 3-12mm (0.1-0.5 in.)
Assay Percent Range 99.9%, metal basis
Molecular Formula SZn
Formula Weight 97.43
MDL Number MFCD00011301
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8829909

  SDS
alfa aesar
8829909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8829918

  SDS
alfa aesar
8829918
50g Each for $45.81

AA8829930

  SDS
alfa aesar
8829930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 97.43
Quantity 10g
CAS 1314-98-3
Physical Form Pieces, 3-12mm (0.1-0.5 in.)
Chemical Name or Material Zinc Sulfide
Molecular Formula SZn
Assay Percent Range 99.9%, metal basis
MDL Number MFCD00011301