Please login to your online account to display your discounted pricing

Zinc Sulfate Solution, 1.180 Specific Gravity, for Concentration of Intestinal Parasites, Eggs, and Cysts, Ricca Chemical

$44.18 - $153.42

Chemical Identifiers

Chemical Name or Material Zinc Sulfate Solution
Name Note 1.180 Specific Gravity
CAS 7732-18-5
CAS Min % 74.0
CAS Max % 77.0
CAS 7446-20-0
CAS Min % 24.31
CAS Max % 25.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

9650-32

 
ricca chemical company
RI965032
Poly natural 1L Each for $65.48

9650-16

 
ricca chemical company
RI965016
Poly natural 500mL Each for $44.18

9650-1

  SDS
ricca chemical company
RI96501
Poly natural 4L Each for $153.42
Description & Specifications

Specifications

Quantity 1L
Solvent or Matrix Aqueous solution
CAS Min % 74.0
CAS Max % 77.0
Chemical Name or Material Zinc Sulfate Solution
Name Note 1.180 Specific Gravity
CAS 7732-18-5
CAS 7446-20-0
Packaging Poly natural
CAS Min % 24.31
CAS Max % 25.3

For Concentration of Intestinal Parasites, Eggs, and Cysts