Learn More
Alfa Aesar™ Zinc sulfate monohydrate, Zn 35.5%
Manufacturer: Alfa Aesar™ 14160A7
Specifications
Zinc sulfate monohydrate | |
H2O5SZn | |
UN3077 | |
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate | |
RNZCSKGULNFAMC-UHFFFAOYSA-L | |
zinc;sulfate;hydrate | |
62639 | |
Coarse Powder | |
5kg |
7446-19-7 | |
MFCD00149893 | |
14,10159 | |
Soluble in water | |
O.[O-]S(=O)(=O)[O-].[Zn+2] | |
179.451 | |
179.45 (161.43 Anhydrous) | |
Zn 35.5% |
Chemical Identifiers
7446-19-7 | |
179.451 | |
RNZCSKGULNFAMC-UHFFFAOYSA-L | |
62639 | |
O.[O-]S(=O)(=O)[O-].[Zn+2] |
H2O5SZn | |
MFCD00149893 | |
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate | |
zinc;sulfate;hydrate |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
P280i-P273-P301+P310-P305+P351+P338-P315-P501a
Wear eye/face protection.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Get immediate medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : TSCA