Please login to your online account to display your discounted pricing

Zinc Sulfate Heptahydrate (USP/FCC), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLZ76500

 View more versions of this product

Catalog No. Z76-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 287.56
Melting Point 100°C
Quantity 500g
Assay 99.0 to 108.7% (FCC)
pH 4.4 to 6
Physical Form Solid
Molecular Formula O4SZn · 7 H2O
Decomposition Information 500°C
Color White
Identification Pass Test (FCC)
Lead (Pb) 0.002% max. (USP)
Residual Solvents Meets Requirements (USP)
Alkalis and Alkaline Earths 0.5% max. (FCC)
Mercury (Hg) 5mg/kg max. (FCC)
Selenium (Se) 0.003% max. (FCC)
Acidity Pass Test (FCC)
Arsenic (As) 14ppm max. (USP)
Cadmium (Cd) 5mg/kg max. (FCC)
Chemical Name or Material Zinc Sulfate Heptahydrate
Packaging Poly Bottle
MDL Number MFCD00149894
ChemAlert Storage Symbol Gray
CAS 7446-20-0
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:0

Specifications

Chemical Name or Material Zinc Sulfate Heptahydrate
CAS 7446-20-0
Purity Grade FCC/USP
Assay 99.0 to 108.7% (FCC)
Molecular Formula O4SZn · 7 H2O
Formula Weight 287.56
MDL Number MFCD00149894
Accessories
 

Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents