missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Sulfate, Heptahydrate, ACS Grade, 99.0 to 103.0%, LabChem™

$92.34 - $92.34

Chemical Identifiers

CAS 7446-20-0
CAS Max % 100
Linear Formula Zn(NO3)2·6H2O
Formula Weight 297.48
Chemical Name or Material Zinc Sulfate, Heptahydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC272201 Lab Chem
LC272201
500g Each for $92.34
Specifications Description & Specifications

Chemical Identifiers

7446-20-0
Zn(NO3)2·6H2O
Zinc Sulfate, Heptahydrate
100
297.48
ACS Grade

Specifications

7446-20-0
Crystals
297.48
Passes Test
Soluble in water
Room Temperature
White
500g
99.0 to 103.0%
100
Zn(NO3)2·6H2O
1,970kg/m3
Poly Bottle
1970kg/m3
Sulfur compounds
100°C
Zinc Sulfate, Heptahydrate
ACS Grade

Safety and Handling

GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Warning