missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Sulfate, Heptahydrate, ACS, 99.5 to 103.0%, MilliporeSigma™

Manufacturer:  MilliporeSigma 1088835000

 View more versions of this product

Catalog No. M1088835000



Specifications

Specifications

Zinc Sulfate, Heptahydrate
7446-20-0
H14O11SZn
ccris 5563
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
287.541
CHEBI:32312
Powder
800 to 1000kg/m3
White
EMSURE™, For Analysis
99.5 to 103.0%
ZnSO4 · 7H2O
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.54
ACS/EP/ISO/Reagent
1.97g/cm3 (20°C)
5kg

Chemical Identifiers

7446-20-0
287.541
ccris 5563
CHEBI:32312
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
H14O11SZn
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

Documents