missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate heptahydrate, 99%, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC424600010

 View more versions of this product

Catalog No. AC424600010



Description

Description

Specifications

Specifications

Zinc sulfate heptahydrate
99.0 to 103% (Complexometry)
ZnSO4·7H2O
10ppm max.
ccris 5563, unii-n57ji2k7wp, white vitriol heptahydrate, zinc sulfate 1:1 heptahydrate, zinc sulfate heptahydrate, zinc sulfate heptahydrate 1:1:7, zinc sulfate jan, zinc sulfate znso4 heptahydrate, zinc vitriol heptahydrate, znso4.7h2o
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.53
99%
50ppm max.
5ppm max.
10ppm max.
50ppm max.
20ppm max.
100ppm max.
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol
100.0°C
1kg
7446-20-0
O4SZn·7H2O
MFCD00149894
15, 10359
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol,insoluble in alcohol
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
287.53
CHEBI:32312
Crystals
ACS Reagent
5ppm max
0.01% max
30ppm max.
3ppm max.
Plastic bottle
500ppm max.
White
4.4 to 6.0 (5% soln. at 25°C)

Chemical Identifiers

7446-20-0
287.53
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
O4SZn·7H2O
MFCD00149894
ccris 5563, unii-n57ji2k7wp, white vitriol heptahydrate, zinc sulfate 1:1 heptahydrate, zinc sulfate heptahydrate, zinc sulfate heptahydrate 1:1:7, zinc sulfate jan, zinc sulfate znso4 heptahydrate, zinc vitriol heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : ZH5300000

SDS Documents