missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate heptahydrate, 99.5%, for analysis, ACROS Organics™

Manufacturer:  Acros Organics AC205980050

 View more versions of this product

Catalog No. AC205980050



Specifications

Specifications

Zinc sulfate heptahydrate
99.5%
ZnSO4·7H2O
15, 10359
1ppm max.
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.53
99.5%
5ppm max.
10ppm max.
0.002% max.
10ppm max.
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol
100.0°C
5kg
7446-20-0
O4SZn·7H2O
MFCD00149894
ccris 5563
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol,insoluble in alcohol
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
287.53
CHEBI:32312
Crystals or Crystalline Powder
Analysis
10ppm max.
3ppm max.
Plastic bottle
10ppm max.
White
4.4 to 6 (5% soln. at 20°C)

Chemical Identifiers

7446-20-0
287.53
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
O4SZn·7H2O
MFCD00149894
ccris 5563
CHEBI:32312
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents