Please login to your online account to display your discounted pricing

Zinc Sulfate, For Nitrogen (Ammonia), Nitrogen (Organic), 10% (w/v), LabChem™

$114.11 - $114.11

Chemical Identifiers

Chemical Name or Material Zinc Sulfate
CAS 7732-18-5
CAS Max % 83.76
CAS 7446-20-0
CAS Max % 16.24
Name Note For Nitrogen (Ammonia), Nitrogen (Organic)
Linear Formula Zn(NO3)2·6H2O
Formula Weight 297.48
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC272302

 
lab chem
LC272302
Poly Bottle 1L Each for $114.11
Description & Specifications

Specifications

Formula Weight 297.48
Chemical Name or Material Zinc Sulfate
Viscosity 1.35cSt
Decomposition Information Sulfur compounds
Color Clear, colorless
Quantity 1L
Name Note For Nitrogen (Ammonia), Nitrogen (Organic)
Physical Form Liquid
Assay 10% (w/v)
Linear Formula Zn(NO3)2·6H2O
Density 1.1g/mL
CAS 7732-18-5
CAS Max % 83.76
Health Hazard 2 GHS H Statement
Toxic to aquatic life.
Identification Passes Test
CAS 7446-20-0
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 16.24
Solubility Soluble in water
Specific Gravity 1.1g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic to aquatic life.