Please login to your online account to display your discounted pricing

Zinc Sulfate, Certified, 1.00M ±0.01M (2.0N), LabChem™

$94.83 - $94.83

Chemical Identifiers

Chemical Name or Material Zinc Sulfate
CAS 7732-18-5
CAS Max % 75.06
CAS 7446-20-0
CAS Max % 24.94
Purity Grade Certified Grade
Linear Formula Zn(NO3)2·6H2O
Formula Weight 297.48
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC272401

 
lab chem
LC272401
500mL Each for $94.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 297.48
Solubility Soluble in water
Concentration 1.00M ±0.01M (2.0N)
Storage Information Room Temperature
Linear Formula Zn(NO3)2·6H2O
Specific Gravity 1.16g/mL
Identification Passes Test
Viscosity 1.66cSt
CAS 7446-20-0
CAS Max % 24.94
CAS 7732-18-5
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 75.06
Density 1.16g/mL
Decomposition Information Sulfur compounds
Packaging Poly Bottle
Chemical Name or Material Zinc Sulfate
Quantity 500mL
Health Hazard 2 GHS H Statement
Toxic to aquatic life.
Physical Form Liquid
Color Clear, colorless
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic to aquatic life.