missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Sulfate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7446-20-0
Physical Form Granular
Molecular Formula ZnSO4·7H2O
Formula Weight 287.54
Chemical Name or Material Zinc Sulfate, 7-Hydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK8880212 Avantor Performance
888006
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK8880500 Avantor Performance
888012
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7446-20-0
ZnSO4·7H2O
Zinc Sulfate, 7-Hydrate
Granular
287.54
ACS Grade

Specifications

287.54
Poly
ZnSO4·7H2O
Zinc Sulfate, 7-Hydrate
2.5kg
7446-20-0
Granular
ACS Grade