Please login to your online account to display your discounted pricing

Zinc Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$129.59 - $630.13

Chemical Identifiers

Chemical Name or Material Zinc Sulfate
CAS 7446-20-0
Name Note 7-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula ZnSO4·7H2O
Formula Weight 287.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004612

 
avantor performance
438204
Poly Bottle 125g Each for $129.59

02004613

 
avantor performance
438205
Poly Bottle 2.5kg Each for $630.13

02004614

 
avantor performance
438201
Poly Bottle 500g Each for $204.07
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 287.56
Chemical Name or Material Zinc Sulfate
CAS 7446-20-0
Molecular Formula ZnSO4·7H2O
Name Note 7-Hydrate
Quantity 125g
Packaging Poly Bottle
Specific Gravity 1L = 1.97kg