missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate, 100mM aq. soln., Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J60963AE

 View more versions of this product

Catalog No. AAJ60963AE



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Zinc sulfate
7733-02-0
100 mL
ZnSO4
UN3082
NWONKYPBYAMBJT-UHFFFAOYSA-L
zinc;sulfate
24424
161.43
Colorless
Liquid
O4SZn
MFCD00011302
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
[Zn++].[O-]S([O-])(=O)=O
161.44
CHEBI:35176
100mM aq. soln.

Chemical Identifiers

7733-02-0
161.44
NWONKYPBYAMBJT-UHFFFAOYSA-L
24424
zinc;sulfate
O4SZn
MFCD00011302
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
CHEBI:35176
[Zn++].[O-]S([O-])(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions