Please login to your online account to display your discounted pricing

Zinc Sulfate, 0.0200 Molar, Aqueous solutions, Ricca Chemicals


Quantity 20L

Manufacturer: ricca chemical company  RIR965700020F

 View more versions of this product

Catalog No. R965700020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Zinc Sulfate
CAS 7446-20-0
CAS 7732-18-5
Physical Form Liquid
Color Clear colorless
Quantity 20L

Specifications

Chemical Name or Material Zinc Sulfate
CAS 7446-20-0
CAS 7732-18-5
Physical Form Liquid