missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc stearate, ACROS Organics™
Manufacturer: Fisher Scientific Acros AC263780010
Specifications
Zinc stearate | |
Pass Test | |
Plastic bottle | |
118.0°C to 128.0°C | |
557-05-1 | |
[CH3(CH2)16CO2]2Zn | |
15, 10358 | |
Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] | |
632.34 | |
632.34 | |
12.5 to 14.0% (ZnO) |
>100°C | |
Authentic | |
White | |
1kg | |
C36H70O4Zn | |
MFCD00013031 | |
coad, hydense, hytech, mathe, metallac, stearates, zinc distearate, zinc octadecanoate, zinc stearate, zink distearat | |
XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
zinc;octadecanoate | |
11178 | |
Powder |
Chemical Identifiers
557-05-1 | |
632.34 | |
XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
11178 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] |
C36H70O4Zn | |
MFCD00013031 | |
coad, hydense, hytech, mathe, metallac, stearates, zinc distearate, zinc octadecanoate, zinc stearate, zink distearat | |
zinc;octadecanoate |
Safety and Handling
EINECSNumber : 209-151-9
RTECSNumber : ZH5200000
TSCA : TSCA