Please login to your online account to display your discounted pricing

Zinc Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

$96.77 - $96.77

Chemical Identifiers

Chemical Name or Material Zinc Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
CAS 010196-18-6
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ICP-030

 
ultra scientific
ICP030
125mL Each for $96.77
Description & Specifications

Specifications

Quantity 125mL
Analyte or Component Names Zinc,water,nitric acid
Solvent or Matrix Water with dilute nitric acid
CAS Max % 97.9
CAS Max % 0.1
CAS 007732-18-5
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 007697-37-2
CAS 010196-18-6
Chemical Name or Material Zinc Standard
CAS Max % 2.0