Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis)

$572.61 - $572.61

Chemical Identifiers

Chemical Name or Material Zinc sputtering target
CAS 7440-66-6
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41658KS

 
alfa aesar™
41658KS
1 each Each for $572.61
Description & Specifications

Specifications

Chemical Name or Material Zinc sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011291
CAS 7440-66-6
Odor Odorless
Quantity 1 each