Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

$503.26 - $503.26

Chemical Identifiers

Chemical Name or Material Zinc sputtering target
CAS 7440-66-6
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41657KS

  SDS
alfa aesar™
41657KS
1 each Each for $503.26
Description & Specifications

Specifications

TSCA TSCA
Quantity 1 each
CAS 7440-66-6
MDL Number MFCD00011291
Assay 99.99%
Odor Odorless
Chemical Name or Material Zinc sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay Percent Notes (metals basis)