Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis)

$485.18 - $485.18

Chemical Identifiers

Chemical Name or Material Zinc sputtering target
CAS 7440-66-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41656KS

  SDS
alfa aesar™
41656KS
1 each Each for $485.18
Description & Specifications

Specifications

TSCA TSCA
Assay 99.99%
Chemical Name or Material Zinc sputtering target
Quantity 1 each
Assay Percent Notes (metals basis)
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
CAS 7440-66-6
Odor Odorless
MDL Number MFCD00011291