Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Magnetron sputtering source

$364.64 - $364.64

Chemical Identifiers

Chemical Name or Material Zinc sputtering target
CAS 7440-66-6
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41655KS

  SDS
alfa aesar™
41655KS
1 each Each for $364.64
Description & Specifications

Specifications

Odor Odorless
Assay Percent Notes (metals basis)
CAS 7440-66-6
Quantity 1 each
MDL Number MFCD00011291
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.99%
Chemical Name or Material Zinc sputtering target
TSCA TSCA