missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc shot, 1-6mm (0.04-0.24in), Puratronic, 99.9999% (metals basis), Alfa Aesar

Manufacturer:  Alfa Aesar 10760A1

 View more versions of this product

Catalog No. AA10760A1



Specifications

Specifications

Zinc
1-6mm (0.04-0.24 in.)
Gray shot
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
Odorless
23994
65.3
Gray
1kg
7440-66-6
99.9999% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Shot, 1 To 6mm (0.04 To 0.24 in.)

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

Documents