missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc selenite

$19.70 - $19.70

Chemical Identifiers

CAS 13597-46-1
Formula Weight 192.33
MDL Number MFCD00011297
UN Number UN2630
Chemical Name or Material Zinc selenite
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1258609 SDS Alfa Aesar™
1258609
10g Each for $19.70
AA1258622 SDS Alfa Aesar™
1258622
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1258636 SDS Alfa Aesar™
1258636
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA12586A3 SDS Alfa Aesar™
12586A3
2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13597-46-1
MFCD00011297
Zinc selenite
192.33
UN2630

Specifications

13597-46-1
192.33
UN2630
Zinc selenite
Powder
MFCD00011297
10g

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

TSCA : TSCA