Learn More
Zinc selenite, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012586A3
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc selenite | |
Powder | |
O3SeZn | |
UN2630 | |
PEUPCBAALXHYHP-UHFFFAOYSA-L | |
zinc;selenite | |
159686 |
13597-46-1 | |
2 kg | |
MFCD00011297 | |
zinc selenite, selenious acid zinc salt 1:1, selenious acid, zinc salt 1:1, zinkselenit, acmc-1bpna, zinc 2+ ion selenite, selenious acid, zinc salt, zinc 2+ selenite, zinc, ion zn2+ selenit | |
[Zn++].[O-][Se]([O-])=O | |
192.35 | |
192.33 |
Chemical Identifiers
13597-46-1 | |
192.35 | |
PEUPCBAALXHYHP-UHFFFAOYSA-L | |
159686 | |
[Zn++].[O-][Se]([O-])=O |
O3SeZn | |
MFCD00011297 | |
zinc selenite, selenious acid zinc salt 1:1, selenious acid, zinc salt 1:1, zinkselenit, acmc-1bpna, zinc 2+ ion selenite, selenious acid, zinc salt, zinc 2+ selenite, zinc, ion zn2+ selenit | |
zinc;selenite |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
EINECSNumber : 237-048-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only