missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc selenite

Manufacturer:  Alfa Aesar™ 1258636

 View more versions of this product

Catalog No. AA1258636



Specifications

Specifications

Zinc selenite
O3SeZn
UN2630
PEUPCBAALXHYHP-UHFFFAOYSA-L
zinc;selenite
159686
Powder
13597-46-1
MFCD00011297
acmc-1bpna
[O-][Se](=O)[O-].[Zn+2]
192.348
192.33
500g

Chemical Identifiers

13597-46-1
192.348
PEUPCBAALXHYHP-UHFFFAOYSA-L
159686
[O-][Se](=O)[O-].[Zn+2]
O3SeZn
MFCD00011297
acmc-1bpna
zinc;selenite
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 237-048-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents