Learn More
Specifications
Zinc selenite | |
O3SeZn | |
UN2630 | |
PEUPCBAALXHYHP-UHFFFAOYSA-L | |
zinc;selenite | |
159686 | |
Powder |
13597-46-1 | |
MFCD00011297 | |
zinc selenite, selenious acid zinc salt 1:1, selenious acid, zinc salt 1:1, zinkselenit, acmc-1bpna, zinc 2+ ion selenite, selenious acid, zinc salt, zinc selenite, zinc 2+ selenite, zinc, ion zn2+ selenit | |
[O-][Se](=O)[O-].[Zn+2] | |
192.348 | |
192.33 | |
100g |
Chemical Identifiers
13597-46-1 | |
192.348 | |
PEUPCBAALXHYHP-UHFFFAOYSA-L | |
159686 | |
[O-][Se](=O)[O-].[Zn+2] |
O3SeZn | |
MFCD00011297 | |
zinc selenite, selenious acid zinc salt 1:1, selenious acid, zinc salt 1:1, zinkselenit, acmc-1bpna, zinc 2+ ion selenite, selenious acid, zinc salt, zinc selenite, zinc 2+ selenite, zinc, ion zn2+ selenit | |
zinc;selenite |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
TSCA : TSCA