missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc selenide, 99.999% (metals basis)

$92.59 - $1,242.16

Chemical Identifiers

CAS 1315-09-9
MDL Number MFCD00011296
Synonym zinc selenide crystal optic disc, 13mm x 1mm, polished both sides
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1099909 View Documents Alfa Aesar™
1099909
10g Each for $92.59
 
AA1099918 View Documents Alfa Aesar™
1099918
50g Each for $337.53
 
AA1099930 View Documents Alfa Aesar™
1099930
250g Each for $1,242.16
 
Specifications

Chemical Identifiers

1315-09-9
MFCD00011296
zinc selenide crystal optic disc, 13mm x 1mm, polished both sides

Specifications

1315-09-9
UN3283
zinc selenide crystal optic disc, 13mm x 1mm, polished both sides
Powder
5.42g/mL at 15°C
Air Sensitive
10g
(metals basis)
MFCD00011296
14,10156
144.33
99.999%
Odorless
>1100°C
Zinc selenide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

TSCA : TSCA