missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc selenide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1099930

 View more versions of this product

Catalog No. AA1099930



Specifications

Specifications

Zinc selenide
(metals basis)
UN3283
zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick, zinc selenide znse, zinc selenide, powder, 10 mum trace metals basis, zinc selenide, spectro, znse
144.33
99.999%
Odorless
>1100°C
1315-09-9
MFCD00011296
14,10156
Insoluble in water.
Powder
5.42 g/mL at 15°C
Air Sensitive
250g

Chemical Identifiers

1315-09-9
MFCD00011296
zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick, zinc selenide znse, zinc selenide, powder, 10 mum trace metals basis, zinc selenide, spectro, znse
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P301+P310-P314-P330-P501c

H301-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 215-259-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents