missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc selenide, 99.99%, (trace metal basis), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC223640100

 View more versions of this product

Catalog No. AC223640100


 

Specifications Description & Specifications

Specifications

144.34
10g
For trace metal basis analysis
99.99%
Glass bottle
MFCD00011296
SeZn
Zinc selenide
Shiny Fine Powder
Yellow
15, 10356
ZnSe
0.01% max.
99.99%
1315-09-9

Chemical Identifiers

1315-09-9
SeZn
144.34
Zinc selenide, 99.99%
99.99%
ZnSe
MFCD00011296
For trace metal basis analysis

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-259-7

TSCA : TSCA