Please login to your online account to display your discounted pricing

Zinc selenide, 99.99%, (trace metal basis), ACROS Organics™

$72.62 - $177.09

Chemical Identifiers

Chemical Name or Material Zinc selenide
Name Note 99.99%
CAS 1315-09-9
Purity Grade For trace metal basis analysis
Assay Percent Range 99.99%
Molecular Formula SeZn
Linear Formula ZnSe
Formula Weight 144.34
MDL Number MFCD00011296
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC223640100

  SDS
thermo fisher scientific
AC223640100
Glass bottle 10g Each for $72.62

AC223640250

  SDS
thermo fisher scientific
223640250
Glass bottle 25g Each for $177.09
Description & Specifications

Specifications

Physical Form Shiny Fine Powder
Formula Weight 144.34
Quantity 10g
Color Yellow
EINECS Number 215-259-7
TSCA TSCA
Purity Grade For trace metal basis analysis
Merck Index 15, 10356
Assay Percent Range 99.99%
Linear Formula ZnSe
Chemical Name or Material Zinc selenide
Name Note 99.99%
Packaging Glass bottle
Molecular Formula SeZn
MDL Number MFCD00011296
CAS 1315-09-9
Total Trace Metal Impurities 0.01% max.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

for ultra trace analysis

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.