missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc rod, 8mm (0.31 in.) dia., 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 44936DT

 View more versions of this product

Catalog No. AA44936DT



Specifications

Specifications

Zinc rod
8mm (0.31 in.) dia.
(metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
99.9999%
250mm
7440-66-6
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
Odorless
23994
Rod

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents