Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc rod, 1.27cm (0.5 in.) dia. x 15cm (5.9 in.) long, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Zinc rod
CAS 7440-66-6
Name Note 1.27cm (0.5 in.) dia. x 15cm (5.9 in.) long
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10434KH

  SDS
alfa aesar™
10434KH
2 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10434KK

  SDS
alfa aesar™
10434KK
10 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
MDL Number MFCD00011291
Physical Form Rod
Assay Percent Notes (metals basis)
Weight ≈9.03g/cm
Name Note 1.27cm (0.5 in.) dia. x 15cm (5.9 in.) long
TSCA TSCA
CAS 7440-66-6
Chemical Name or Material Zinc rod
Quantity 2 pieces
Assay 99.5%