Please login to your online account to display your discounted pricing

Zinc, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSZ13100

 View more versions of this product

Catalog No. SZ13-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg
Viscosity [NULL]
Density 1.1g/cm³
CAS 7697-37-2
Traceability to NIST LOT NUMBER USED ZINC REF. STANDARD
CAS 7732-18-5
Chemical Name or Material Zinc, Reference Standard Solution
MDL Number MFCD00011291
Packaging Poly Bottle
Name Note 1000ppm ±1%
Concentration 990 to 1010ppm
ChemAlert Storage Symbol White
CAS 1314-13-2

Specifications

Chemical Name or Material Zinc, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7697-37-2
CAS 1314-13-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011291
Accessories
 

SDS & Safety Documents