Please login to your online account to display your discounted pricing

Zinc, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$75.41 - $105.82

Chemical Identifiers

Chemical Name or Material Zinc, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7697-37-2
CAS 1314-13-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SZ13-500

  SDS
fisher scientific company
SZ13500
Poly Bottle 500mL Each for $105.82
Case of 6 Each for $634.92

SZ13-100

  SDS
fisher scientific company
FLSZ13100
Poly Bottle 100mL Each for $75.41
Case of 6 Each for $452.46
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
Vapor Pressure 14mmHg
Density 1.1g/cm³
CAS 7697-37-2
CAS 7732-18-5
Packaging Poly Bottle
CAS 1314-13-2
Chemical Name or Material Zinc, Reference Standard Solution
MDL Number MFCD00011291
Concentration 990 to 1010ppm
Traceability to NIST LOT NUMBER USED ZINC REF. STANDARD
ChemAlert Storage Symbol White
Name Note 1000ppm ±1%
Safety and Handling