Please login to your online account to display your discounted pricing

Zinc protoporphyrin IX, Tocris

Heme oxygenase and guanylyl cyclase inhibitor

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material Zinc protoporphyrin IX
Assay Percent Range >98%
Molecular Formula C34H32N4O4Zn
Formula Weight 626.03
Synonym 7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid, zinc complex
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

074610

 
r & d systems
074610
10mg Each for $139.08

074650

 
r & d systems
074650
50mg Each for $585.69
Description & Specifications

Specifications

Chemical Name or Material Zinc protoporphyrin IX
CAS 15442-64-5
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C34H32N4O4Zn
Formula Weight 626.03
Synonym 7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid, zinc complex
Quantity 10mg
Safety and Handling

Store at Room Temperature