missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc protoporphyrin, 96%

Manufacturer:  Alfa Aesar™ 32068MF

 View more versions of this product

Catalog No. AA32068MF



Specifications

Specifications

Zinc protoporphyrin
96%
MFCD00011612
50mg
Protoporphyrin IX zinc(II)
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]
626.034
626.04
15442-64-5
C34H32N4O4Zn
Purple to Red
Odorless
FUTVBRXUIKZACV-UHFFFAOYSA-L
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
131664383
Crystalline

Chemical Identifiers

15442-64-5
626.034
FUTVBRXUIKZACV-UHFFFAOYSA-L
131664383
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]
C34H32N4O4Zn
MFCD00011612
Protoporphyrin IX zinc(II)
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
Safety and Handling

Safety and Handling

RTECSNumber : ZG9210300

TSCA : TSCA

SDS Documents