Learn More
Zinc protoporphyrin, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 032068MA
Description
Zinc protoporphyrin-9 is a potent, selective heme oxygenase inhibitor, interleukin-1 antagonist, GCS inhibitor, and NOS1, NOS2, and NOS3 inhibitor. In olfactory neuronal cultures, this product has been used to block CO biosynthesis, which lowered cGMP concentrations. Zinc protoporphyrin-9 is an extremely useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc protoporphyrin | |
Purple to Red | |
C34H32N4O4Zn | |
10 mg | |
Insoluble in water. Soluble in ethanol,and DMSO,pyridine,or dimethyl formamide. | |
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2] | |
626.034 | |
626.04 | |
Crystalline |
15442-64-5 | |
Odorless | |
MFCD00011612 | |
zinc protoporphyrin | |
FUTVBRXUIKZACV-UHFFFAOYSA-L | |
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate | |
131664383 | |
96% |
Chemical Identifiers
15442-64-5 | |
626.034 | |
FUTVBRXUIKZACV-UHFFFAOYSA-L | |
131664383 | |
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2] |
C34H32N4O4Zn | |
MFCD00011612 | |
zinc protoporphyrin | |
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate |
Safety and Handling
EINECSNumber : 239-455-7
RTECSNumber : ZG9210300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only