missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13835AP

 View more versions of this product

Catalog No. AA13835AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Zinc
Liquid
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Liquid
Colorless
7440-66-6,7440-66-6,7732-18-5
Zn
Nitric acid: 5.0%; Zinc: 0.1%; Water: 94.9%
[Zn]
65.38
CHEBI:30185
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
CHEBI:30185
[Zn]
Zn
MFCD00011291
23994
zinc
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

Documents