missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc phthalocyanine, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 422750050

 View more versions of this product

Catalog No. AC422750050



Description

Description

Specifications

Specifications

Zinc phthalocyanine
5% max. (105°C)
Purple
98%
96%
phthalocyanine zinc, phthalocyanine, zinc, zinc ii phthalocyanine, zinc phthalocyanine, zinc phthalocyanine, dye content 97 %, znpc
PODBBOVVOGJETB-UHFFFAOYSA-N
577.91
CHEBI:51218
Powder
Authentic
Glass bottle
5g
14320-04-8
C32H16N8Zn
Solubility in water: insoluble
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
2735172
577.91
96%

Chemical Identifiers

14320-04-8
577.91
phthalocyanine zinc, phthalocyanine, zinc, zinc ii phthalocyanine, zinc phthalocyanine, zinc phthalocyanine, dye content 97 %, znpc
CHEBI:51218
C32H16N8Zn
PODBBOVVOGJETB-UHFFFAOYSA-N
2735172
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
Safety and Handling

Safety and Handling

EINECSNumber : 238-262-5

SDS Documents