Please login to your online account to display your discounted pricing

Zinc phthalocyanine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  422750250

 View more versions of this product

Catalog No. AC422750250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 577.91
Physical Form Powder
Quantity 25g
Color Purple
Packaging Glass bottle
Assay Percent Range 96%
Chemical Name or Material Zinc phthalocyanine
Molecular Formula C32H16N8Zn
Name Note 98%
Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
UV_Vis 95% min.
CAS 14320-04-8

Specifications

Chemical Name or Material Zinc phthalocyanine
Name Note 98%
CAS 14320-04-8
Assay Percent Range 96%
Molecular Formula C32H16N8Zn
Formula Weight 577.91