missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc phthalocyanine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03955309

 View more versions of this product

Catalog No. AA3955309



Description

Description

Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc phthalocyanine
Purple
C32H16N8Zn
10 g
Insoluble in water.
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
2735172
577.91
Powder
14320-04-8
Odorless
MFCD00041955
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
PODBBOVVOGJETB-UHFFFAOYSA-N
577.916
CHEBI:51218
95%

Chemical Identifiers

14320-04-8
577.916
PODBBOVVOGJETB-UHFFFAOYSA-N
2735172
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
C32H16N8Zn
MFCD00041955
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
CHEBI:51218
Safety and Handling

Safety and Handling

EINECSNumber : 238-262-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only