missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01301314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc phosphate (ortho) | |
White | |
25 g | |
O8P2Zn3 | |
MFCD00036282 | |
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2] | |
386.08 | |
386.05 | |
Puratronic™ | |
3.75 g/mL |
7779-90-0 | |
Powder | |
(metals basis) | |
Zn3(PO4)2 | |
UN3077 | |
LRXTYHSAJDENHV-UHFFFAOYSA-H | |
trizinc;diphosphate | |
24519 | |
99.995% | |
Odorless |
Chemical Identifiers
7779-90-0 | |
386.08 | |
LRXTYHSAJDENHV-UHFFFAOYSA-H | |
24519 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2] |
O8P2Zn3 | |
MFCD00036282 | |
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc | |
trizinc;diphosphate |
Safety and Handling
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 231-944-3
RTECSNumber : TD0590000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only