missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1301306

 View more versions of this product

Catalog No. AA1301306



Specifications

Specifications

Zinc phosphate (ortho)
(metals basis)
Zn3(PO4)2
UN3077
LRXTYHSAJDENHV-UHFFFAOYSA-H
trizinc;diphosphate
24519
Powder
Puratronic™
Odorless
5g
7779-90-0
O8P2Zn3
MFCD00036282
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
386.08
386.05
99.995%
3.75g/mL
White

Chemical Identifiers

7779-90-0
386.08
LRXTYHSAJDENHV-UHFFFAOYSA-H
24519
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
O8P2Zn3
MFCD00036282
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
trizinc;diphosphate
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : TD0590000

TSCA : TSCA

Documents